Assay Details
Target Gene Details
Entrez Gene ID: | 233905 |
Gene Name: | zinc finger protein 646 |
Gene Aliases: |
6820429M01 |
Location: |
Chr.7:127877036-127885996 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp646 | NM_172749.4 | 1 | 204 | NP_766337.2 |
BC079907.1 | 1 | 204 | AAH79907.1 |
More Information
Set Membership: |
Intragenic Exonic |