Assay Details
Target Gene Details
Entrez Gene ID: | 11808 |
Gene Name: | apolipoprotein A-IV |
Gene Aliases: |
Apoa-4 |
Location: |
Chr.9:46240844-46243458 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apoa4 | NM_007468.2 | 1 | 58 | NP_031494.2 |
BF660098.1 | 1 | 70 |
More Information
Set Membership: |
Intragenic Exonic |