Assay Details
Target Gene Details
Entrez Gene ID: | 78252 |
Gene Name: | neurexophilin and PC-esterase domain family, member 2 |
Gene Aliases: |
4432416J03Rik, AI323362, Fam55b |
Location: |
Chr.9:48318003-48353453 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nxpe2 | NM_030069.3 | 6 | 1776 | NP_084345.2 |
AK019469.1 | 6 | 1676 | BAB31740.1 | |
AK033667.1 | 6 | 1541 | BAC28418.1 | |
AK157096.1 | 6 | 1714 | BAE33960.1 | |
AK162300.1 | 6 | 1538 | ||
AK165213.1 | 5 | 946 | BAE38080.1 | |
BC140955.1 | 6 | 1659 |
Target Gene Details
Entrez Gene ID: | 244853 |
Gene Name: | neurexophilin and PC-esterase domain family, member 4 |
Gene Aliases: |
C130036J11, D930028F11Rik, Fam55d |
Location: |
Chr.9:48162001-48400025 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nxpe4 | AK048132.1 | BAC33253.1 | ||
BC094249.1 | AAH94249.1 |