Assay Details
Target Gene Details
Entrez Gene ID: | 23886 |
Gene Name: | growth differentiation factor 15 |
Gene Aliases: |
MIC-1, NAG-1, SBF |
Location: |
Chr.8:70629393-70632469 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gdf15 | NM_011819.2 | 1 | 262 | NP_035949.2 |
AF159571.1 | 1 | 732 | AAD41410.1 | |
AK144199.1 | 2 | 561 | BAE25763.1 | |
AK159242.1 | 1 | 263 | BAE34924.1 | |
AK160927.1 | 1 | 175 | BAE36095.1 | |
BC067248.1 | 1 | 251 | AAH67248.1 |
More Information
Set Membership: |
Intragenic Exonic |