Assay Details
Target Gene Details
Entrez Gene ID: | 100048658 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 43 |
Gene Aliases: |
ENSMUSG00000070291, OTTMUSG00000019690 |
Location: |
Chr.9:78395777-78423589 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx43 | NM_001191044.1 | 1 | 60 | NP_001177973.1 |