Assay Details
Target Gene Details
Entrez Gene ID: | 321008 |
Gene Name: | RIKEN cDNA 6330408A02 gene |
Gene Aliases: |
- |
Location: |
Chr.7:13258967-13278721 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
6330408A02Rik | NM_177312.4 | 4 | 2015 | NP_796286.2 |
AK018141.1 | 4 | 1775 | BAB31091.1 | |
AK160064.1 | 3 | 2654 | BAE35600.1 | |
BC075681.1 | 3 | 1989 | AAH75681.1 |