Assay Details
Target Gene Details
Entrez Gene ID: | 321008 |
Gene Name: | RIKEN cDNA 6330408A02 gene |
Gene Aliases: |
- |
Location: |
Chr.7:13258967-13278721 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
6330408A02Rik | NM_177312.4 | 3 | 522 | NP_796286.2 |
AK018141.1 | BAB31091.1 | |||
AK160064.1 | 2 | 1161 | BAE35600.1 | |
AK162673.1 | 3 | 493 | BAE37016.1 | |
BC075681.1 | 2 | 496 | AAH75681.1 | |
CV783961.1 | 3 | 523 |