Assay Details
Target Gene Details
Entrez Gene ID: | 233876 |
Gene Name: | HIRA interacting protein 3 |
Gene Aliases: |
B130036O03, C86302 |
Location: |
Chr.7:126861972-126865377 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hirip3 | NM_172746.3 | 4 | 1679 | NP_766334.1 |
AK045122.1 | 4 | 1214 | BAC32232.1 | |
AK135526.1 | 4 | 1222 | ||
AK161657.1 | 4 | 1214 | ||
AK162096.1 | 1 | 241 | BAE36722.1 | |
BC055687.1 | 4 | 1679 | AAH55687.1 |