Assay Details
Target Gene Details
Entrez Gene ID: | 14132 |
Gene Name: | Fc receptor, IgG, alpha chain transporter |
Gene Aliases: |
FcRn |
Location: |
Chr.7:45092992-45103846 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcgrt | NM_010189.3 | 5 | 1172 | NP_034319.2 |
AK146933.1 | 5 | 1137 | ||
AK152879.1 | 5 | 1108 | BAE31564.1 | |
AK180969.1 | ||||
AK212167.1 | ||||
BC003786.1 | 5 | 1107 | AAH03786.1 | |
D37874.1 | 5 | 1105 | BAA07111.1 | |
L17022.1 | 4 | 811 | AAA16904.1 |
More Information
Set Membership: |
Intragenic Exonic |