Assay Details
Target Gene Details
Entrez Gene ID: | 12518 |
Gene Name: | CD79A antigen (immunoglobulin-associated alpha) |
Gene Aliases: |
Ig-alpha, Iga, Igalpha, Ly-54, Ly54, mb-1 |
Location: |
Chr.7:24897511-24902197 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd79a | NM_007655.3 | 2 | 131 | NP_031681.2 |
AK143605.1 | 2 | 132 | BAE25458.1 | |
BC027633.1 | 2 | 128 | AAH27633.1 | |
X13450.1 | 2 | 129 | CAA31801.1 |
More Information
Set Membership: |
Intragenic Exonic |