Assay Details
Target Gene Details
Entrez Gene ID: | 21835 |
Gene Name: | thyroid hormone responsive |
Gene Aliases: |
S14 |
Location: |
Chr.7:97412957-97417510 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Thrsp | NM_009381.2 | 1 | 245 | NP_033407.1 |
AK050300.1 | 1 | 255 | BAC34175.1 | |
AK077300.1 | BAC36737.1 | |||
BC009165.1 | 1 | 245 | AAH09165.1 |