Assay Details
Target Gene Details
Entrez Gene ID: | 259277 |
Gene Name: | kallikrein related-peptidase 8 |
Gene Aliases: |
BSP1, Nrpn, Prss19 |
Location: |
Chr.7:43797552-43803822 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klk8 | NM_008940.2 | 3 | 756 | NP_032966.1 |
AB074296.1 | BAB92021.1 | |||
BC055895.1 | 3 | 270 | AAH55895.1 | |
D30785.1 | 3 | 756 | BAA06451.1 |
More Information
Set Membership: |
Intragenic Exonic |