Assay Details
Target Gene Details
Entrez Gene ID: | 11870 |
Gene Name: | ADP-ribosyltransferase 1 |
Gene Aliases: |
ADPRT, ARTC1, Yac-1 |
Location: |
Chr.7:102101706-102111758 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Art1 | NM_009710.4 | 3 | 790 | NP_033840.2 |
AK003107.1 | 3 | 791 | BAB22570.1 | |
AK009284.1 | BAB26194.1 | |||
BC063760.1 | 3 | 771 | AAH63760.1 | |
BC132492.1 | 3 | 778 | ||
BC132496.1 | 3 | 778 | ||
X95842.1 | 2 | 752 | CAA65099.1 |
More Information
Set Membership: |
Intragenic Exonic |