Assay Details
Target Gene Details
Entrez Gene ID: | 192658 |
Gene Name: | ret finger protein-like 4 |
Gene Aliases: |
D7Ertd486e, Rfpl4a |
Location: |
Chr.7:5109787-5116911 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rfpl4 | NM_001145013.1 | NP_001138485.1 | ||
NM_138954.3 | 1 | 182 | NP_620404.1 | |
AK136022.1 | BAE22777.1 | |||
AK139569.1 | 1 | 184 | ||
AY070253.1 | 1 | 172 | AAL55432.1 | |
BC069834.1 | AAH69834.1 |