Assay Details
Target Gene Details
Entrez Gene ID: | 56538 |
Gene Name: | kallikrein related-peptidase 11 |
Gene Aliases: |
2310015I08Rik, Prss20, TLSP |
Location: |
Chr.7:43774617-43779263 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klk11 | NM_001177373.1 | 6 | 974 | NP_001170844.1 |
NM_019974.2 | 5 | 940 | NP_064358.1 | |
AB016226.1 | 6 | 824 | BAA88825.1 | |
AB016227.1 | 6 | 934 | BAA36955.1 | |
AK009360.1 | 6 | 975 | BAB26241.2 | |
AK009451.1 | 2 | 265 | BAB26297.1 | |
AK009659.1 | 6 | 949 | ||
AK009720.1 | 6 | 975 | BAB26461.2 | |
AK019102.1 | 2 | 265 | BAB31548.1 | |
BC148310.2 | 5 | 649 |
More Information
Set Membership: |
Intragenic Exonic |