Assay Details
Target Gene Details
Entrez Gene ID: | 52377 |
Gene Name: | reticulocalbin 3, EF-hand calcium binding domain |
Gene Aliases: |
6030455P07Rik, D530026G20Rik, D7Ertd671e, RLP49 |
Location: |
Chr.7:45082914-45092257 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rcn3 | NM_026555.2 | 7 | 1302 | NP_080831.2 |
AK003918.1 | 6 | 972 | BAB23076.1 | |
AK077943.1 | 7 | 1242 | BAC37077.1 | |
AK082762.1 | 7 | 1232 | BAC38608.1 | |
AK086705.1 | 7 | 1243 | ||
AK132143.1 | 7 | 1236 | ||
AV129283.1 | ||||
BC005487.1 | 6 | 1139 | AAH05487.1 | |
BC025602.1 | 7 | 1253 | AAH25602.1 | |
BC055903.1 | 7 | 1249 | AAH55903.1 | |
BC092069.1 | 7 | 1200 | AAH92069.1 |
More Information
Set Membership: |
Intragenic Exonic |