Assay Details
Target Gene Details
Entrez Gene ID: | 622320 |
Gene Name: | potassium channel tetramerisation domain containing 21 |
Gene Aliases: |
EG622320 |
Location: |
Chr.7:97332311-97350216 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kctd21 | NM_001039039.3 | 2 | 536 | NP_001034128.2 |
AK141547.1 | 2 | 533 | BAE24730.1 | |
BC139363.1 | 1 | 485 | ||
BC139365.1 | 1 | 485 | ||
CX221410.1 |
More Information
Set Membership: |
Intragenic Exonic |