Assay Details
Target Gene Details
Entrez Gene ID: | 13871 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 2 |
Gene Aliases: |
AA407812, AU020867, AW240756, CXPD, Ercc-2, XPD |
Location: |
Chr.7:19382039-19395692 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ercc2 | NM_007949.4 | 23 | 3325 | NP_031975.2 |
AK004652.1 | 19 | 2977 | BAB23443.1 | |
AK082323.1 | 16 | 2579 | ||
AK082761.1 | 23 | 3326 | BAC38607.1 | |
AK141692.1 | 23 | 3343 | ||
AK145969.1 | 23 | 3334 | BAE26794.1 | |
BC034517.1 | 22 | 3261 | AAH34517.1 |
Target Gene Details
Entrez Gene ID: | 232943 |
Gene Name: | kinesin light chain 3 |
Gene Aliases: |
BC017147, Klct |
Location: |
Chr.7:19394440-19404442 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klc3 | NM_001286038.1 | NP_001272967.1 | ||
NM_001286039.1 | NP_001272968.1 | |||
NM_146182.4 | NP_666294.1 | |||
AK158094.1 | BAE34354.1 | |||
BC017147.1 | AAH17147.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |