Assay Details
Target Gene Details
Entrez Gene ID: | 66985 |
Gene Name: | Ras association (RalGDS/AF-6) domain family (N-terminal) member 7 |
Gene Aliases: |
2400009B11Rik, AW210608 |
Location: |
Chr.7:141215560-141218658 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rassf7 | NM_025886.3 | 6 | 1398 | NP_080162.3 |
AK002243.1 | 6 | 1337 | BAB21960.1 | |
AK010305.1 | 6 | 1424 | ||
AW990538.1 | 1 | 133 | ||
BC011131.1 | 5 | 1325 | AAH11131.1 |
More Information
Set Membership: |
Intragenic Exonic |