Assay Details
Target Gene Details
Entrez Gene ID: | 66985 |
Gene Name: | Ras association (RalGDS/AF-6) domain family (N-terminal) member 7 |
Gene Aliases: |
2400009B11Rik, AW210608 |
Location: |
Chr.7:141215560-141218658 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rassf7 | NM_025886.3 | 3 | 452 | NP_080162.3 |
AK002243.1 | 3 | 392 | BAB21960.1 | |
AK010305.1 | 3 | 479 | ||
BC011131.1 | 2 | 379 | AAH11131.1 | |
CK617449.1 | 3 | 452 |
More Information
Set Membership: |
Intragenic Exonic |