Assay Details
Target Gene Details
Entrez Gene ID: | 17188 |
Gene Name: | MYC-associated zinc finger protein (purine-binding transcription factor) |
Gene Aliases: |
PUR1, Pur-1, SAF-1, SAF-2 |
Location: |
Chr.7:127022123-127026553 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Maz | NM_010772.1 | 5 | 1687 | NP_034902.1 |
BC004758.1 | 8 | 1454 | ||
DQ150586.1 | 4 | 1401 |
More Information
Set Membership: |
Intragenic Exonic |