Assay Details
Target Gene Details
Entrez Gene ID: | 67164 |
Gene Name: | lipoyl(octanoyl) transferase 2 (putative) |
Gene Aliases: |
2610209A20Rik |
Location: |
Chr.7:100159277-100160931 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lipt2 | NM_026010.2 | 2 | 592 | NP_080286.2 |
AK008917.1 | 2 | 573 | BAB25967.1 | |
AK011925.1 | 2 | 592 | BAB27920.1 | |
AK013127.1 | 2 | 574 | ||
BC115472.1 | 2 | 592 | ||
BC115473.1 | 2 | 587 |
More Information
Set Membership: |
Intragenic Exonic |