Assay Details
Target Gene Details
Entrez Gene ID: | 22793 |
Gene Name: | zyxin |
Gene Aliases: |
9530098H06Rik, R75157 |
Location: |
Chr.6:42349621-42360213 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zyx | NM_001289617.1 | 5 | 1130 | NP_001276546.1 |
NM_001289618.1 | 4 | 1037 | NP_001276547.1 | |
NM_001289619.1 | 4 | 1037 | NP_001276548.1 | |
NM_011777.3 | 5 | 1130 | NP_035907.1 | |
AK031415.1 | 5 | 928 | BAC27394.1 | |
AK035743.1 | 5 | 901 | BAC29173.1 | |
AK076491.1 | 5 | 900 | ||
AK090110.1 | 5 | 897 | ||
AK147812.1 | 5 | 1131 | BAE28154.1 | |
AK161709.1 | 4 | 833 | ||
AK165290.1 | 3 | 971 | ||
AK170571.1 | 5 | 904 | BAE41886.1 | |
AK176971.1 | ||||
AK184760.1 | ||||
AK184932.1 | ||||
AK190613.1 | ||||
BC054775.1 | 4 | 996 | AAH54775.1 | |
X99063.1 | 5 | 1088 | CAA67510.1 | |
Y07711.1 | 4 | 838 | CAA68984.1 |
More Information
Set Membership: |
Intragenic Exonic |