Assay Details
Target Gene Details
Entrez Gene ID: | 75690 |
Gene Name: | ZV-set and immunoglobulin domain containing 10 like |
Gene Aliases: |
- |
Location: |
Chr.7:43463061-43472023 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vsig10l | NM_001290316.1 | 3 | 703 | NP_001277245.1 |