Assay Details
Target Gene Details
Entrez Gene ID: | 68197 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2 |
Gene Aliases: |
1810004I06Rik, 2010300P09Rik, G1, R75094 |
Location: |
Chr.7:97400003-97407800 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufc2 | NM_024220.2 | 1 | 169 | NP_077182.1 |
AI413456.1 | ||||
AK002373.1 | 1 | 77 | BAB22050.1 | |
AK003380.2 | 1 | 72 | ||
AK008502.1 | BAB25703.1 | |||
AK008889.1 | 1 | 77 | BAB25955.1 | |
AK019002.1 | 1 | 96 | BAB31504.1 | |
AK153547.1 | 1 | 99 | BAE32084.1 | |
AK166594.1 | 1 | 96 | BAE38880.1 | |
AK168342.1 | 1 | 121 | BAE40279.1 | |
BC002097.1 | 1 | 92 | AAH02097.1 | |
CK128102.1 | 1 | 169 |
More Information
Set Membership: |
Intragenic Exonic |