Assay Details
Target Gene Details
Entrez Gene ID: | 269854 |
Gene Name: | N-acetyltransferase 14 |
Gene Aliases: |
BC047219 |
Location: |
Chr.7:4918888-4925006 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nat14 | NM_201355.3 | 3 | 1244 | NP_958743.1 |
AK078272.1 | BAC37199.1 | |||
AK158627.1 | 3 | 1245 | BAE34587.1 | |
AK163549.1 | 3 | 972 | ||
BC047219.1 | 3 | 880 | AAH47219.1 |
More Information
Set Membership: |
Intragenic Exonic |