Assay Details
Target Gene Details
Entrez Gene ID: | 17872 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 15A |
Gene Aliases: |
9630030H21, Gadd34, Myd116 |
Location: |
Chr.7:45522917-45526268 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r15a | NM_008654.2 | 3 | 1990 | NP_032680.1 |
AK136336.1 | 3 | 1946 | ||
AK154697.1 | 2 | 2472 | BAE32770.1 | |
AK167317.1 | 4 | 1715 | BAE39419.1 | |
AW493042.1 | 1 | 356 | ||
BC138485.1 | 3 | 1855 | ||
BC138486.1 | 3 | 1855 | ||
X51829.1 | 3 | 1935 | CAA36128.1 |