Assay Details
Target Gene Details
Entrez Gene ID: | 330657 |
Gene Name: | protease, serine 53 |
Gene Aliases: |
Pol3s, mFLJ00289 |
Location: |
Chr.7:127885444-127890970 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss53 | NM_001081268.1 | 11 | 1839 | NP_001074737.1 |
AK220244.1 | 11 | 1796 | BAD90169.1 | |
BC039632.1 | 8 | 1409 | AAH39632.1 | |
BC147509.1 | 11 | 1836 | ||
BC151010.1 | 11 | 1836 | ||
BC151017.1 | 11 | 1836 | ||
BC172059.1 | 10 | 1619 |
More Information
Set Membership: |
Intragenic Exonic |