Assay Details
Target Gene Details
Entrez Gene ID: | 12337 |
Gene Name: | calpain 5 |
Gene Aliases: |
nCL-3 |
Location: |
Chr.7:98121559-98179298 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Capn5 | NM_001301250.1 | NP_001288179.1 | ||
NM_007602.4 | NP_031628.1 | |||
AK155179.1 | BAE33097.1 | |||
AK164293.1 | BAE37722.1 | |||
AK170034.1 | BAE41522.1 | |||
BC014767.1 | AAH14767.1 | |||
U85020.1 | AAD00559.1 | |||
Y10656.1 | CAA71666.1 |
Target Gene Details
Entrez Gene ID: | 18378 |
Gene Name: | olfactory marker protein |
Gene Aliases: |
- |
Location: |
Chr.7:98143359-98145447 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Omp | NM_011010.2 | 1 | 1045 | NP_035140.1 |
AK133724.1 | 1 | 1076 | BAE21801.1 | |
BC046600.1 | 1 | 1045 | AAH46600.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |