Assay Details
Target Gene Details
Entrez Gene ID: | 12406 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade H, member 1 |
Gene Aliases: |
BERF-1, Cbp1, Cbp2, Hsp47, J6, Serpinh2, gp46 |
Location: |
Chr.7:99345375-99353239 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpinh1 | NM_001111043.1 | 1 | 125 | NP_001104513.1 |
NM_001111044.1 | 1 | 125 | NP_001104514.1 | |
NM_001285776.1 | 1 | 125 | NP_001272705.1 | |
NM_009825.2 | 1 | 125 | NP_033955.2 | |
AK029080.1 | BAC26283.1 | |||
AK031457.1 | BAC27413.1 | |||
AK076474.1 | BAC36360.1 | |||
AK077660.1 | BAC36935.1 | |||
AK077937.1 | BAC37075.1 | |||
AK077992.1 | ||||
AK079460.1 | BAC37652.1 | |||
AK086519.1 | BAC39683.1 | |||
AK086523.1 | BAC39684.1 | |||
AK089993.1 | BAC41032.1 | |||
AK090103.1 | BAC41093.1 | |||
AK159697.1 | BAE35297.1 | |||
AK159998.1 | BAE35546.1 | |||
AK166000.1 | BAE38510.1 | |||
AK167403.1 | BAE39492.1 | |||
AK168442.1 | BAE40347.1 | |||
AK168870.1 | BAE40689.1 | |||
AK169422.1 | ||||
BC085143.1 | AAH85143.1 | |||
CA751137.1 | 1 | 174 | ||
J05609.1 | AAA03200.1 | |||
X60676.1 | CAA43091.1 |
More Information
Set Membership: |
Intragenic Exonic |