Assay Details
Target Gene Details
Entrez Gene ID: | 56546 |
Gene Name: | secretory blood group 1 |
Gene Aliases: |
Fut10, Fut3 |
Location: |
Chr.7:45677686-45694402 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec1 | NM_001271578.1 | 4 | 1539 | NP_001258507.1 |
AF113532.1 | 1 | 942 | AAD25351.1 | |
AK030246.1 | 4 | 1542 | BAC26862.1 | |
BC100713.1 | 1 | 941 | AAI00714.1 | |
BC103557.1 | 1 | 941 | AAI03558.1 | |
BC103558.1 | 1 | 941 | AAI03559.1 | |
BC103559.1 | 1 | 941 | AAI03560.1 | |
BC115947.1 | 1 | 941 |
More Information
Set Membership: |
Intragenic Exonic |