Assay Details
Target Gene Details
Entrez Gene ID: | 245884 |
Gene Name: | family with sequence similarity 71, member F2 |
Gene Aliases: |
AI507611, GARI |
Location: |
Chr.6:29277539-29290684 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam71f2 | NM_001101486.1 | 5 | 1063 | NP_001094956.1 |
BC151115.1 | 5 | 1030 | ||
BC151117.1 | 5 | 1030 |