Assay Details
Target Gene Details
Entrez Gene ID: | 231452 |
Gene Name: | SDA1 domain containing 1 |
Gene Aliases: |
4931421J16, AA591032, AW538460 |
Location: |
Chr.5:92284010-92310024 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdad1 | NM_172713.2 | 22 | 2643 | NP_766301.1 |
AK029881.1 | 22 | 2645 | BAC26657.1 | |
AK084789.1 |