Assay Details
Target Gene Details
Entrez Gene ID: | 19227 |
Gene Name: | parathyroid hormone-like peptide |
Gene Aliases: |
PTH-like, Pthrp |
Location: |
Chr.6:147252110-147264085 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pthlh | NM_008970.3 | 3 | 771 | NP_032996.2 |
AJ278119.1 | 2 | 266 | CAC39218.1 | |
AY183377.1 | 2 | 285 | AAO25537.1 | |
AY220497.1 | 3 | 532 | AAO64343.1 | |
BC058187.1 | 3 | 576 | AAH58187.1 | |
CB194603.1 |
More Information
Set Membership: |
Intragenic Exonic |