Assay Details
Target Gene Details
Entrez Gene ID: | 232023 |
Gene Name: | vesicular, overexpressed in cancer, prosurvival protein 1 |
Gene Aliases: |
AW146242, Ecop |
Location: |
Chr.6:57752264-57825125 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vopp1 | NM_146168.1 | 5 | 2485 | NP_666280.1 |
AK030975.1 | 5 | 2472 | BAC27198.1 | |
AK031812.1 | 5 | 2490 | BAC27562.1 | |
AK170152.1 | 5 | 2436 | BAE41599.1 | |
AK170832.1 | 5 | 2436 | BAE42060.1 | |
BC024822.1 | 5 | 2485 | AAH24822.1 |