Assay Details
Target Gene Details
Entrez Gene ID: | 22223 |
Gene Name: | ubiquitin carboxy-terminal hydrolase L1 |
Gene Aliases: |
AW822034, C88048, PGP 9.5, PGP9.5, R75593, UCH-L1, UCHL-1, gad |
Location: |
Chr.5:66676121-66687234 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uchl1 | NM_011670.2 | 9 | 821 | NP_035800.2 |
AB025313.1 | 9 | 672 | BAA84083.1 | |
AF172334.1 | 9 | 660 | AAD51029.1 | |
AK004066.1 | 9 | 698 | ||
AK013729.1 | 9 | 701 | BAB28976.1 | |
AK170728.1 | 7 | 659 | BAE41984.1 | |
BC039177.1 | 9 | 687 | AAH39177.1 |
More Information
Set Membership: |
Intragenic Exonic |