Assay Details
Target Gene Details
Entrez Gene ID: | 20751 |
Gene Name: | sepiapterin reductase |
Gene Aliases: |
AA409688, ENSMUSG00000071345, Gm10328 |
Location: |
Chr.6:85133680-85137764 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spr | NM_011467.2 | 3 | 822 | NP_035597.2 |
AK004941.1 | 3 | 632 | ||
AK207238.1 | 2 | 227 | ||
AK216452.1 | 2 | 227 | ||
BC010400.1 | 3 | 819 | AAH10400.1 | |
BC054423.1 | 3 | 806 | AAH54423.1 | |
BY590544.1 | 1 | 62 | ||
S71375.1 | AAP19647.1 | |||
S77493.1 | 3 | 824 | AAB33611.1 | |
W65601.1 | 2 | 281 | ||
Z21947.1 | CAA79942.1 |
More Information
Set Membership: |
Intragenic Exonic |