Assay Details
Target Gene Details
Entrez Gene ID: | 140491 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3A |
Gene Aliases: |
GM, RG1, RGL |
Location: |
Chr.6:14713822-14755274 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r3a | NM_080464.2 | 1 | 405 | NP_536712.2 |
AK084518.1 | 1 | 406 | BAC39208.2 | |
AK084719.1 | 1 | 236 | BAC39262.2 | |
BC109007.1 | 1 | 377 | AAI09008.1 |
More Information
Set Membership: |
Intragenic Exonic |