Assay Details
Target Gene Details
Entrez Gene ID: | 12123 |
Gene Name: | harakiri, BCL2 interacting protein (contains only BH3 domain) |
Gene Aliases: |
AI838259, Bid3, DP5, harakiri |
Location: |
Chr.5:118169764-118189478 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hrk | NM_007545.2 | 2 | 2028 | NP_031571.1 |