Assay Details
Target Gene Details
Entrez Gene ID: | 77042 |
Gene Name: | hyaluronoglucosaminidase 4 |
Gene Aliases: |
4632428M18Rik, CSHY |
Location: |
Chr.6:24748367-24766519 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hyal4 | NM_029848.1 | 5 | 1538 | NP_084124.1 |
AB524072.1 | 4 | 1401 | ||
BC125402.1 | 5 | 1538 | ||
BC132096.1 | 5 | 1538 |
More Information
Set Membership: |
Intragenic Exonic |