Assay Details
Target Gene Details
Entrez Gene ID: | 666311 |
Gene Name: | zinc finger and SCAN domain containing 25 |
Gene Aliases: |
EG666311, Zfp498 |
Location: |
Chr.5:145280468-145291495 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zscan25 | NM_001081431.1 | 1 | 150 | NP_001074900.1 |
BC150878.1 | 1 | 141 | ||
BC151110.1 | 1 | 141 |
More Information
Set Membership: |
Intragenic Exonic |