Assay Details
Target Gene Details
Entrez Gene ID: | 12386 |
Gene Name: | catenin (cadherin associated protein), alpha 2 |
Gene Aliases: |
AI481747, Catna, Catna2, cdf, chp |
Location: |
Chr.6:76881637-77979694 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctnna2 | NM_001109764.1 | NP_001103234.1 | ||
NM_009819.2 | NP_033949.2 | |||
NM_145732.2 | NP_663785.2 | |||
AK158916.1 | BAE34726.1 | |||
BC079648.1 | AAH79648.1 | |||
D25281.1 | BAA04969.1 | |||
D25282.1 | BAA04970.1 |
Target Gene Details
Entrez Gene ID: | 74342 |
Gene Name: | leucine rich repeat transmembrane neuronal 1 |
Gene Aliases: |
4632401D06Rik, AW125451 |
Location: |
Chr.6:77242480-77259915 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrrtm1 | NM_028880.3 | 1 | 422 | NP_083156.2 |
AK014545.1 | 1 | 421 | ||
AK053345.1 | 1 | 411 | BAC35354.1 | |
AK053364.1 | BAC35363.1 | |||
AK080836.1 | 1 | 451 | ||
BC027803.1 | 1 | 304 | AAH27803.1 | |
BY724290.1 | 1 | 423 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |