Assay Details
Target Gene Details
Entrez Gene ID: | 66627 |
Gene Name: | 2-oxoglutarate and iron-dependent oxygenase domain containing 2 |
Gene Aliases: |
1300006G11Rik, 5730405M13Rik, AU041636, AW552395 |
Location: |
Chr.5:124112058-124115480 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ogfod2 | NM_025671.2 | 7 | 1291 | NP_079947.1 |
AK004943.1 | 7 | 1292 | BAB23687.1 | |
AK017497.1 | 7 | 1291 | BAC25517.1 | |
AK018534.1 | 7 | 1292 | BAB31259.1 | |
AK168946.1 | 7 | 1209 | ||
BC022762.1 | 6 | 1139 | AAH22762.1 |
More Information
Set Membership: |
Intragenic Exonic |