Assay Details
Target Gene Details
Entrez Gene ID: | 66627 |
Gene Name: | 2-oxoglutarate and iron-dependent oxygenase domain containing 2 |
Gene Aliases: |
1300006G11Rik, 5730405M13Rik, AU041636, AW552395 |
Location: |
Chr.5:124112058-124115480 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ogfod2 | NM_025671.2 | 6 | 565 | NP_079947.1 |
AK004916.1 | 3 | 1228 | ||
AK004943.1 | 6 | 566 | BAB23687.1 | |
AK017497.1 | 6 | 566 | BAC25517.1 | |
AK018534.1 | 6 | 566 | BAB31259.1 | |
AK168946.1 | ||||
BC022762.1 | 5 | 413 | AAH22762.1 | |
BC069872.1 | 6 | 568 | AAH69872.1 |
More Information
Set Membership: |
Intragenic Exonic |