Assay Details
Target Gene Details
Entrez Gene ID: | 12034 |
Gene Name: | prohibitin 2 |
Gene Aliases: |
AU044498, BAP, Bap37, Bcap37, REA |
Location: |
Chr.6:124712289-124716945 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phb2 | NM_007531.2 | 1 | 137 | NP_031557.2 |
AK002334.1 | 1 | 105 | ||
AK011107.1 | 1 | 105 | ||
AK090088.1 | 1 | 92 | BAE43403.1 | |
AK132052.1 | 1 | 99 | BAE20962.1 | |
AK132058.1 | 1 | 126 | BAE20964.1 | |
AK135729.1 | 1 | 55 | ||
AK151952.1 | 1 | 131 | BAE30825.1 | |
AK152375.1 | 1 | 127 | ||
AK153368.1 | 1 | 127 | ||
AK158423.1 | 1 | 142 | ||
AY211613.1 | 1 | 99 | AAP47231.1 | |
AY319418.1 | AAP86652.1 | |||
BB595613.1 | ||||
BC145875.1 | 1 | 89 | ||
X78683.1 | 1 | 96 | CAA55350.1 |