Assay Details
Target Gene Details
Entrez Gene ID: | 320127 |
Gene Name: | diacylglycerol kinase, iota |
Gene Aliases: |
C130010K08Rik |
Location: |
Chr.6:36837381-37300060 on Build GRCm38 |
Assay Gene Location: | Within Exon 34 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dgki | NM_001081206.1 | 34 | 4108 | NP_001074675.1 |
AK139313.1 | 3 | 1105 |
More Information
Set Membership: |
Intragenic Exonic |