Assay Details
Target Gene Details
Entrez Gene ID: | 213819 |
Gene Name: | CAS1 domain containing 1 |
Gene Aliases: |
Cas1, Cast1 |
Location: |
Chr.6:4600563-4643381 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Casd1 | NM_145398.2 | 9 | 1017 | NP_663373.2 |
AB091829.1 | 9 | 1017 | BAC77246.1 | |
AK202913.1 | 1 | 52 | ||
AK217605.1 | 1 | 51 | ||
BC058953.1 | AAH58953.1 | |||
BC125377.1 | 9 | 940 | ||
BC125379.1 | 9 | 940 |
More Information
Set Membership: |
Intragenic Exonic |