Assay Details
Target Gene Details
Entrez Gene ID: | 11628 |
Gene Name: | activation-induced cytidine deaminase |
Gene Aliases: |
Aid, Arp2 |
Location: |
Chr.6:122553809-122564180 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aicda | NM_009645.2 | 3 | 474 | NP_033775.1 |
AF132979.1 | 3 | 473 | AAD41793.1 | |
AF529837.1 | 2 | 381 | AAM95424.1 | |
AK080144.1 | 2 | 473 |
More Information
Set Membership: |
Intragenic Exonic |