Assay Details
Target Gene Details
Entrez Gene ID: | 101202 |
Gene Name: | HEPACAM family member 2 |
Gene Aliases: |
AI987662 |
Location: |
Chr.6:3457089-3498189 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hepacam2 | NM_178899.5 | 3 | 502 | NP_849230.1 |
AK033728.1 | 3 | 504 | BAC28450.1 | |
AK050501.1 | 3 | 503 | BAC34294.1 | |
BC096374.1 | 3 | 489 | AAH96374.1 |