Assay Details
Target Gene Details
Entrez Gene ID: | 209086 |
Gene Name: | sterile alpha motif domain containing 9-like |
Gene Aliases: |
AA175286, ESTM25, mKIAA2005 |
Location: |
Chr.6:3372257-3399571 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Samd9l | NM_010156.3 | 2 | 1145 | NP_034286.2 |
AK052535.1 | 2 | 1146 | ||
AK172649.1 | 2 | 1033 | ||
AK173329.1 | 2 | 1013 | BAD32607.1 |